missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, Light Weight, MP Biomedicals

Catalog Number p-323371
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
8042-47-5
C16H10N2Na2O7S2
452.363
MFCD00131611
AEOVEGJBKQQFOP-DDVLFWKVSA-L
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
9566064
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
8042-47-5
C16H10N2Na2O7S2
452.363
MFCD00131611
AEOVEGJBKQQFOP-DDVLFWKVSA-L
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
9566064
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'