Få mere at vide
Miltirone, TRC
Artikelnummer. 30482415
Skift visning
Mængde:
2.5 mg
5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30482415 5 mg -
30475876 10 mg -
30468190 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30482415 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3445005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27210-57-7
Molekylær formel C19 H22 O2
Molekylvægt (g/mol) 282.38
IUPAC navn 8,8-dimethyl-2-propan-2-yl-6,7-dihydro-5H-phenanthrene-3,4-dione
SMIL CC(C)C1=Cc2ccc3c(CCCC3(C)C)c2C(=O)C1=O
Tekniske data
Kemisk navn eller materiale Miltirone
Mængde 5 mg
InChI formel InChI=1S/C19H22O2/c1-11(2)14-10-12-7-8-15-13(6-5-9-19(15,3)4)16(12)18(21)17(14)20/h7-8,10-11H,5-6,9H2,1-4H3
Formel vægt 282.162