Beskrivelse
- Dermatologically tested and fresh-scented foam hand soap.
- A gentle soap that helps prevent dry skin with moisturising, lipid-replenishing ingredients and a pearly shine.
- 96% of ingredients are from natural origin.
- Minimise energy use - this soap is proven effective in cold water (based on testing at 20 degrees Celcius).
Tekniske data
Tekniske data
| Produkttype | Hand Soap |
| Farve | Clear |
| Form | Skum |
| Emballage | Plastic Bottle |
| Mængde | 8/Pk. |
| Størrelse | 525 mL |