Få mere at vide
Milciclib, MedChemExpress
Artikelnummer. 16480317
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16480317 50 mg
16464976 100 mg
16423968 10 mg
16406687 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16480317 Leverandør Medchem Express Leverandørnr. HY10424
MedChemExpress Milciclib (PHA-848125) is a potent, ATP-competitive and dual inhibitor of CDK and Tropomyosin receptor kinase (TRK), with IC50s of 45, 150, 160, 363, 398 nM and 53 nM for cyclin A/CDK2, cyclin H/CDK7, cyclin D1/CDK4, cyclin E/CDK2, cyclin B/CDK1 and TRKA, respectively.
Milciclib (PHA-848125) is a potent, ATP-competitive and dual inhibitor of CDK and Tropomyosin receptor kinase (TRK), with IC50s of 45, 150, 160, 363, 398 nM and 53 nM for cyclin A/CDK2, cyclin H/CDK7, cyclin D1/CDK4, cyclin E/CDK2, cyclin B/CDK1 and TRKA, respectively.
Tekniske data
Kemisk navn eller materiale Milciclib
Fysisk form Solid
CAS 802539-81-7
Farve Yellow
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C25H32N8O
Til brug med (applikation) Cancer-Kinase/protease
Synonym PHA-848125
Opløselighedsinformation DMSO : 20 mg/mL (43.42 mM; Need ultrasonic)
SMIL O=C(NC)C1=NN(C2=C1C(C)(CC3=CN=C(N=C23)NC4=CC=C(C=C4)N5CCN(CC5)C)C)C
Molekylvægt (g/mol) 460.57
Formel vægt 460.57
Procent renhed 99.44%