missing translation for 'onlineSavingsMsg'
Få mere at vide

Mikimopine, TRC

Artikelnummer. 30480321
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480321 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30480321 Leverandør Toronto Research Chemicals Leverandørnr. TRCM38550810MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Mikimopine
CAS 120293-52-9
Molekylær formel C11H13N3O6
Mængde 10 mg
Synonym (4R,6S)-4-(2-Carboxyethyl)-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine-4,6-dicarboxylic Acid
InChI formel InChI=1S/C11H13N3O6/c15-7(16)1-2-11(10(19)20)8-5(12-4-13-8)3-6(14-11)9(17)18/h4,6,14H,1-3H2,(H,12,13)(H,15,16)(H,17,18)(H,19,20)/t6-,11+/m0/s1
SMIL OC(=O)CC[C@]1(N[C@@H](Cc2[nH]cnc12)C(=O)O)C(=O)O
IUPAC navn (4R,6S)-4-(2-carboxyethyl)-1,5,6,7-tetrahydroimidazo[4,5-c]pyridine-4,6-dicarboxylic acid
Molekylvægt (g/mol) 283.24
Formel vægt 283.0804
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.