Få mere at vide
Miglustat hydrochloride, MedChemExpress
Artikelnummer. 16487268
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16487268 10 mg
16479787 5 mg
16453566 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16487268 Leverandør Medchem Express Leverandørnr. HY17020A
MedChemExpress Miglustat hydrochloride is an inhibitor of glucosylceramide synthase, primarily to treat Type I Gaucher disease (GD1).
Miglustat hydrochloride is an inhibitor of glucosylceramide synthase, primarily to treat Type I Gaucher disease (GD1).
Tekniske data
Kemisk navn eller materiale Miglustat hydrochloride
Fysisk form Oil
CAS 210110-90-0
Farve Light Brown
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C10H22ClNO4
Til brug med (applikation) Neuroscience-Neuromodulation
Synonym N-Butyldeoxynojirimycin hydrochlorideNB-DNJ hydrochlorideOGT 918 hydrochloride
Opløselighedsinformation DMSO : 65 mg/mL (254.16 mM; Need ultrasonic) ∣H2O : ≥ 34 mg/mL (132.95 mM)
SMIL O[C@H]1[C@H](O)[C@@H](CO)N(CCCC)C[C@@H]1O.[H]Cl
Molekylvægt (g/mol) 255.74
Formel vægt 255.74
Procent renhed 98.0%