Få mere at vide
Miglitol, TRC
Artikelnummer. 30132868
Skift visning
Mængde:
10 mg
25 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132868 25 mg
30138651 1 g
17958674 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132868 Leverandør Toronto Research Chemicals Leverandørnr. TRCM34420025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72432-03-2
Molekylær formel C8 H17 N O5
Molekylvægt (g/mol) 207.22
Synonym 3,4,5-Piperidinetriol, 1-(2-hydroxyethyl)-2-(hydroxymethyl)-, (2R,3R,4R,5S)- (9CI, ACI), (2R,3R,4R,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)-3,4,5-piperidinetriol (ACI), 3,4,5-Piperidinetriol, 1-(2-hydroxyethyl)-2-(hydroxymethyl)-, [2R-(2α,3β,4α,5β)]- (ZCI), (2R,3R,4R,5S)-1-(2-Hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol, BAY 1099, BAY-m 1099, Diastabol, Glyset, Miglitol, Miglitose, N-(2-Hydroethyl)deoxynojirimycin, N-(2-Hydroxyethyl)-1-deoxynojirimycin, N-(2-Hydroxyethyl)moranoline, Plumarol, Seibule
IUPAC navn (2R,3R,4R,5S)-1-(2-hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol
SMIL OCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO
Tekniske data
Kemisk navn eller materiale Miglitol
Mængde 25 mg
InChI formel InChI=1S/C8H17NO5/c10-2-1-9-3-6(12)8(14)7(13)5(9)4-11/h5-8,10-14H,1-4H2/t5-,6+,7-,8-/m1/s1
Formel vægt 207.1107