Learn More
Miele Professional™ Miele Process Documentation
APH 301 Connection cable type 1 for connection of a machine with serial interface for a PC/Net500.
Brand: Miele Professional™ 7951420
412.30 DKK valid until 2024-12-31
Use promo code "23282" to get your promotional price.
Additional Details : Weight : 0.14000kg
Description
- Compatible with Miele G7831, G 788x ,G 789x, and PS 120xB
- Compatible with numerous appliances from Melag, Sirona, W and H and Hawo
- Cable length 3 m
Specifications
Process/Operating Documentation/Communication | |
Washer-disinfectors, laboratory |