missing translation for 'onlineSavingsMsg'
Få mere at vide

Midecamycin, 95%, BLD Pharm™

Artikelnummer. 30963609
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
250 mg
1 g
5 g
25 g
Pakningsstørrelse:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
30963609 5 g 5g
30960968 1 g 1g
30954495 100 mg 100mg
30960741 250 mg 250mg
30953396 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30963609 Leverandør BLD Pharm Leverandørnr. BD1401255g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Specifications

Kemisk navn eller materiale Midecamycin
CAS 35457-80-8
Molekylær formel C41H67NO15
MDL nummer MFCD00865000
Mængde 5 g
SMIL O[C@H]1[C@](O[C@H](C)[C@@H](O[C@@](O[C@@H](C)[C@@H]2OC(CC)=O)([H])C[C@]2(O)C)[C@@H]1N(C)C)([H])O[C@@H]([C@H](C[C@H]([C@H](/C=C/C=C/C3)O)C)CC=O)[C@H]([C@](CC(O[C@@H]3C)=O)([H])OC(CC)=O)OC
Molekylvægt (g/mol) 813.97
Procent renhed 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.