Få mere at vide
Thermo Scientific™ Microtiter Rør og stativer
Shop alle MBP Molecular BioProducts produkter
Produkttype:
Frostet mikrotiterrør
Microtiter Rør
Sterilitet:
Ikke steril
Ikke-steril
Steril
Format:
Bulk
Massevis
Robotstativ
Pakningsstørrelse:
Pakke med 1000
Pakke med 10000
Pakke med 960
Beskrivelse
Forsegl mikrotiterrør tæt til opbevaring med polyethylen Thermo Scientific™ Microtiter Rørprop Strips— til brug med 1,2 mL MCT Titertube racks. 1,2 ml mikrotiterrør tilbyder yderligere volumenkapacitet til prøvefortynding. Disse er tilgængelige i et præ-racket 8 x 12 format.
Microtiter Tubes and Caps
- Polyethylene plug strip seals the Microtiter tubes tightly for storage
- Eight plug strips can be easily cut apart for individual samples
- 1.2mL Microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
- Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
- Microtiter tube system is available in bulk packaging or with a SBS standard footprint
- Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
- Rigorous testing to ensure sample integrity and security
Packaging Options
- Microtiter tubes and caps are available sterile and nonsterile
Tekniske data
Tekniske data
| Certificeringer/compliance | Certified free of RNase, DNase, DNA and pyrogens |
| Farve | Natural |
| Volumen (metrisk) | 1.2 mL |
| Mængde | 96 tubes/rack 10 racks/unit, 5 units case |
| Lukningsmateriale | Polyethylene (Plug) |
| Produktlinje | Thermo Scientific |
| Sterilitet | Ikke-steril |
| Format | Robotstativ |
| Produkttype | Frostet mikrotiterrør |