Learn More
Meiji Techno Microscope Stage Micrometer, 1 mm/0.01 mm Scale
Make quick and accurate measurements of your specimen with the Microscope Stage Micrometer.
Brand:  Meiji Techno 48404-80
1937.05 DKK valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Additional Details : Weight : 0.02000kg
Product Code. 15350703
  • 2255.00 DKK / Each
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Description
Description
Product Type: Microscope Stage Micrometer; Scale: 1mm/0.01mm
  • Easily count or measure sample when observing samples under a microscope.
  • Microscope micrometers, also referred to as reticles, are commonly used for measuring or counting specimens.
  • Eyepiece micrometers are small glass discs with markings on them.
  • The micrometer is mounted in one of the two eyepieces and superimposes an image of the markings over the image of the specimen.
  • Stage micrometers are slides with markings of a known dimension that, when viewed under the microscope, can be used to calibrate eyepiece micrometers.
Specifications
Specifications
Microscope Stage Micrometer
1 Ea.