Få mere at vide
Greiner Bio-One Microlon™ Polystyren 96-brønd ELISA Mikroplader
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Tracebility from production through a defined LOT number system
- Manufactured from crystal clear polystyrene
- The footprint is compatible with automated systems
- Well to well spacing : 9 mm
- Alphanumeric well coding
- Suitable for transmission measurements
- Specifically treated to provide an incresed protein binding
- Well profile: F-bottom / standard (flat bottom)
Specifications
Specifications
| Bindende Ejendom | Mellem indbinding |
| Format | Standard |
| Låg | Without Lid |
| Array | 8 x 12 |
| Farve | Clear |
| Materiale | Polystyrene |
| Antal Wells | 96 |
| Sterilitet | Non-sterile |
| Overfladebehandling | Mikrolon 600 |
| Well Shape | Flat Bottom (F-well) |
| Show More |