missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Micro-Filter Funnel, According to Pregl

Micro-Filter Funnel for small volumes.

Brand:  ROBU VitraPOR™ 28040

Additional Details : Weight : 0.06460kg

 View more versions of this product

Product Code. 16853076

  • 1140.00 DKK / Pack of 5
Estimated Shipment: 14-05-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

4 mL
Borosilicate Glass 3.3
0
5/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers