missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Miconazole nitrate salt, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Type | (±)-Miconazole Nitrate Salt |
Infrared Spectrum | Authentic |
Loss on Drying | 0.5% max. (105°C, 2 h) |
Solubility Information | Solubility in water: slightly soluble. |
Formula Weight | 479.14 |
Percent Purity | ≥97.5% (HPLC) |
Quantity | 1 g |
Chemical Name or Material | (±)-Miconazole nitrate salt |
Safety and Handling
Product Identifier
- (±)-Miconazole nitrate salt
- Warning
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'