Få mere at vide
Miconazole EP Impurity G (as Nitrate), Mikromol™
Beskrivelse
Miconazole EP Impurity G (as Nitrate)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-[(2RS)-2-[(2,5-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate |
| CAS | 909277-72-1 |
| Analyt- eller komponentnavne | 1-[(2RS)-2-[(2,5-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H14 Cl4 N2 O . H N O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-[(2RS)-2-[(2,5-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. G (EP) as Nitrate, Miconazole Impurity G as Nitrate, Miconazole Nitrate Impurity G as Nitrate, Miconazole Impurity G as Nitrate |
| InChI formel | InChI=1S/C18H14Cl4N2O.HNO3/c19-13-2-4-16(21)12(7-13)10-25-18(9-24-6-5-23-11-24)15-3-1-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4) |
| SMIL | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3cc(Cl)ccc3Cl)c(Cl)c1 |
| IUPAC navn | 1-[2-(2,4-dichlorophenyl)-2-[(2,5-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid |
| Vis mere |