Få mere at vide
(±)-Miconazole-d5 Nitrate (2,4-dichlorobenzyloxy-d5), TRC
Artikelnummer. 30469792
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30469792 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469792 Leverandør Toronto Research Chemicals Leverandørnr. TRCM34250110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Miconazole-d5 Nitrate (2,4-dichlorobenzyloxy-d5)
CAS 1398065-80-9
Molekylær formel C18 D5 H9 Cl4 N2 O . H N O3
Mængde 10 mg
Synonym (±)-Miconazole-d5 nitrate (2,4-dichlorobenzyloxy-d5), Miconazole-d5 nitrate, Miconazole-D5 (2,4-dichlorobenzyloxy-D5) nitrate, 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl-d3)methoxy-d2]ethyl]imidazole mononitrate, 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorobenzloxy-d5)ethyl]-1H-imidazole nitrate
InChI formel InChI=1S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4)/i1D,2D,7D,10D2;
SMIL [2H]c1c([2H])c(c(Cl)c([2H])c1Cl)C([2H])([2H])OC(Cn2ccnc2)c3ccc(Cl)cc3Cl.O[N+](=O)[O-]
IUPAC navn 1-[2-(2,4-dichlorophenyl)-2-[dideuterio-(2,4-dichloro-3,5,6-trideuteriophenyl)methoxy]ethyl]imidazole;nitric acid
Molekylvægt (g/mol) 484.172
Formel vægt 482.013