Få mere at vide
MGK 11, TRC
Artikelnummer. 30179969
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179969 50 mg
30179739 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179969 Leverandør Toronto Research Chemicals Leverandørnr. TRCM34098050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 126-15-8
Molekylær formel C13 H16 O2
Molekylvægt (g/mol) 204.26
Synonym 4a(4H)-Dibenzofurancarboxaldehyde, 1,5a,6,9,9a,9b-hexahydro- (6CI, 7CI, 8CI, 9CI, ACI), 1,5a,6,9,9a,9b-Hexahydro-4a(4H)-dibenzofurancarboxaldehyde (ACI), 2,3,4,5-Bis(2-butenylene)tetrahydrofurfural, 2,3,4,5-Bis(δ2-butenylene)tetrahydrofurfural, 2,3:4,5-Bis(2-butene-1,4-diyl)tetrahydrofurfural, 2-Furancarboxaldehyde, 2,3:4,5-bis(2-butene-1,4-diyl)tetrahydro-, 4a-Formyl-1,4,4a,5a,6,9,9a,9b-octahydrodibenzofuran, Bisbutenylenetetrahydrofurfural, Dibutylene tetrafurfural, ENT 17596, MGK Repellent 11, MGK-R 11, MKG 11, NSC 52965, R 11, Repel 111
IUPAC navn 4,5a,6,9,9a,9b-hexahydro-1H-dibenzofuran-4a-carbaldehyde
SMIL O=CC12CC=CCC1C3CC=CCC3O2
Tekniske data
Kemisk navn eller materiale MGK 11
Mængde 50 mg
InChI formel InChI=1S/C13H16O2/c14-9-13-8-4-3-6-11(13)10-5-1-2-7-12(10)15-13/h1-4,9-12H,5-8H2
Formel vægt 204.115