missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Abnova™ MGC50811 Recombinant Protein
missing translation for 'orderingAttributeHoverText'
Mængde:
10 μg
25 μg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
| Adgangsnummer | AAH52750 |
| Til brug med (applikation) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulering | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Gen-id (Entrez) | 375307 |
| Molekylvægt (g/mol) | 68.31 |
| Navn | MGC50811 (Human) Recombinant Protein (P01) |
| pH-område | 8 |
| Fremstillingsmetode | In vitro wheat germ expression system |
| Oprensningsmetode | Glutathione Sepharose 4 Fast Flow |
| Kvalitetskontrol test | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction