missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ MGC35295 Recombinant Protein Product Code.: 16179403

Abnova™ MGC35295 Recombinant Protein

Product Code. 16179403
25 μg, 25µg
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16179403

Brand: Abnova™ H00219995P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH31610.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 219995
Molecular Weight (g/mol) 41.91
Name MGC35295 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Source Wheat Germ (in vitro)
Immunogen MVRRGHVGIFFIEGGVPFWGGACFIISGSLSVAAEKNHTSCLVRSSLGTNILSVMAAFAGTAILLMDFGVTNRDVDRGYLAVLTIFTVLEFFTAVIAMHFGCQAIHAQASAPVIFLPNAFSADFNIPSPAASAPPAYDNVAYAQGVV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FLJ34527/MGC35295
Common Name MS4A15
Gene Symbol MS4A15
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ MGC35295 Recombinant Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.