Få mere at vide
Mexiletine-d6 Hydrochloride, TRC
Artikelnummer. 30179832
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179832 1 mg
30179880 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179832 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3408021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1329835-60-0
Molekylær formel C11H12D6ClNO
Molekylvægt (g/mol) 221.76
Synonym 1-(2,6-Dimethylphenoxy)-2-propanamine-d6 Hydrochloride, 1-Methyl-2-(2,6-xylyloxy)ethylamine-d6 Hydrochloride, Katen-d6, Ko 1173-d6, Ko 1173Cl-d6, Mexilitene-d6, Mexitil-d6, Ritalmex-d6;
IUPAC navn 1,1,1,2,3,3-hexadeuterio-3-(2,6-dimethylphenoxy)propan-2-amine;hydrochloride
SMIL Cl.[2H]C([2H])([2H])C([2H])(N)C([2H])([2H])Oc1c(C)cccc1C
Tekniske data
Kemisk navn eller materiale Mexiletine-d6 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C11H17NO.ClH/c1-8-5-4-6-9(2)11(8)13-7-10(3)12;/h4-6,10H,7,12H2,1-3H3;1H/i3D3,7D2,10D;
Formel vægt 221.1454