Få mere at vide
(±)-Mevalonolactone-4,4,5,5-d4, CDN
Artikelnummer. 30312954
Skift visning
Mængde:
0.025 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312954 0.05 g
30312169 0.025 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312954 Leverandør CDN Leverandørnr. CDND41300.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Mevalonolactone-4,4,5,5-d4
CAS 349553-98-6
Molekylær formel C6 D4 H6 O3
Mængde 0.05 g
Synonym 2 H-Pyran-2-one-5,6-d2, tetrahydro-5,6-d2-4-hydroxy-4-methyl- (9 CI), Tetrahydro-4-hydroxy-4-methyl-2 H-pyran-2-one-5,5,6,6-d4, (±)-Mevalolactone-d4, (±)-Mevalonolactone-d4, DL-Mevalonic acid lactone-d4, (±)-Mevalolactone-4,4,5,5-D4, (±)-Mevalolactone-4,4,5,5-D4, (±)-Mevalolactone-4,4,5,5-D4, (±)-Mevalolactone-4,4,5,5-D4
InChI formel InChI=1 S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i2D2,3D2
SMIL [2 H]C1([2 H])OC(=O)CC(C)(O)C1([2 H])[2 H]
IUPAC navn 5,5,6,6-tetradeuterio-4-hydroxy-4-methyloxan-2-one
Molekylvægt (g/mol) 134.17
Formel vægt 134.0881 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity