Få mere at vide
(±)-Mevalonolactone-4,4,5,5,6,6,6-d7, TRC
Artikelnummer. 30472948
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30472948 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472948 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3390260.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (±)-Mevalolactone-D7 (3-methyl-D3,4,4,5,5-D4)
CAS 347840-19-1
Molekylær formel C6H3D7O3
Mængde 0.25 mg
Synonym 2H-Pyran-2-one-5,6-d2, tetrahydro-5,6-d2-4-hydroxy-4-(methyl-d3)- (9CI), Tetrahydro-4-hydroxy-4-(methyl-d3)-2H-pyran-2-one-5,5,6,6-d4, (±)-Mevalolactone-d7, (±)-Mevalonolactone-d7, DL-Mevalonic acid lactone-d7, DL-Mevalonic acid Δ-lactone-d7, DL-Mevalonol
InChI formel InChI=1S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i1D3,2D2,3D2
SMIL O=C1OC([2H])([2H])C([2H])([2H])C(O)(C([2H])([2H])[2H])C1
IUPAC navn 4-hydroxy-4-(methyl-d3)tetrahydro-2H-pyran-2-one-5,5,6,6-d4
Molekylvægt (g/mol) 137.19
Formel vægt 137.11