Beskrivelse
- pH, Redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, ISE, salinity, TDS, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- *Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
- Preventative maintenance and service available through METTLER TOLEDO Service
Specifications
Specifications
| Inkluderer | S400 pH-måler; InLab Expert Pro-ISM elektrode; elektrodeholder; 2 bufferposer til pH 4,01, 7, 9,21 og 10,01; EasyDirect pH-aktiveringsnøgle (pH datastyringssoftware); test certifikat; Vejledning til pH-måling |
| Nøjagtighed (temperatur) | ±0,1 °C |
| Nøjagtighed (mV) | ±0,1 |
| Nøjagtighed (pH) | ±0,002 |
| Udgange | USB, RS-232, Ethernet |
| Område (pH) | -2 til 20 |
| Opløsning (pH) | 0.001, 0.01, 0.1 |
| Kalibreringspunkter (pH) | Maks. 5 punkter, 8 foruddefinerede og 20 brugerdefinerede buffergrupper |
| Rækkevidde (mV) | -2000 til 2000 |
| Opløsning (mV) | 0.1/1mV |
| Show More |