missing translation for 'onlineSavingsMsg'
Få mere at vide

Metsulfuron-methyl-d3, TRC

Artikelnummer. 30481539
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481539 50 mg
30479652 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481539 Leverandør Toronto Research Chemicals Leverandørnr. TRCM22774250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Metsulfuron-methyl D3 (triazine methoxy D3)
CAS 2377723-88-9
Molekylær formel C14 2H3 H12 N5 O6 S
Mængde 50 mg
Synonym Metsulfuron-methyl D3 (triazine methoxy D3), N-[(2-Methoxycarbonyl)phenyl]sulfonyl-N'-(6-trideuteriomethoxy-4-methyl-2-triazinyl)urea, 2-[[[[(4-Trideuteriomethoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]benzoic acid methyl ester
InChI formel InChI=1S/C14H15N5O6S/c1-8-15-12(18-14(16-8)25-3)17-13(21)19-26(22,23)10-7-5-4-6-9(10)11(20)24-2/h4-7H,1-3H3,(H2,15,16,17,18,19,21)/i3D3
SMIL [2H]C([2H])([2H])Oc1nc(C)nc(NC(=O)NS(=O)(=O)c2ccccc2C(=O)OC)n1
IUPAC navn methyl 2-[[4-methyl-6-(trideuteriomethoxy)-1,3,5-triazin-2-yl]carbamoylsulfamoyl]benzoate
Molekylvægt (g/mol) 384.38
Formel vægt 384.0931
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.