Få mere at vide
Metronidazole-d3, TRC
Artikelnummer. 30480544
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30480544 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480544 Leverandør Toronto Research Chemicals Leverandørnr. TRCM97880010MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metronidazole methyl-d3
CAS 83413-09-6
Renhedsgrad noter HPLC
Molekylær formel C6 D3 H6 N3 O3
Mængde 10 mg
Synonym 2-(Methyl-d3)-5-nitro-1H-imidazole-1-ethanol
InChI formel InChI=1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3/i1D3
SMIL [2H]C([2H])([2H])c1ncc([N+](=O)[O-])n1CCO
IUPAC navn 2-[5-nitro-2-(trideuteriomethyl)imidazol-1-yl]ethanol
Molekylvægt (g/mol) 174.17
Formel vægt 174.0832
Procent renhed >95