Få mere at vide
Metronidazole Acetate, TRC
Artikelnummer. 30466454
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466454 10 mg
30465370 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30466454 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33888510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13182-82-6
Molekylær formel C8H11N3O4
Molekylvægt (g/mol) 213.19
Synonym Flubendazole Impurity B, Flubendazole Imp. B (EP), L-Methionine Sulfone
IUPAC navn 2-(2-methyl-5-nitroimidazol-1-yl)ethyl acetate
SMIL CC(=O)OCCn1c(C)ncc1[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale Metronidazole Acetate
Mængde 10 mg
InChI formel InChI=1S/C8H11N3O4/c1-6-9-5-8(11(13)14)10(6)3-4-15-7(2)12/h5H,3-4H2,1-2H3
Formel vægt 213.075