Learn More
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge, Stackable
Shelf ledges may be stacked on top of each other to create taller ledges, Ledges snap onto shelves and posts for secure fit with no tools
Brand:  Metro™ L24N4C
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 11721059
  • 211.00 DKK / Each
Estimated Shipment: 11-09-2024
to see stock.
Add to basket
Description
Description
  • 4 in. high (10cm)
  • Choice of plated or stainless steel finish
Order Info
Posts and shelves sold separately.
Specifications
Specifications
24 in.
61 cm
Metro™ Super Erecta™ wire shelving
10 cm
4 in.
Chrome