Learn More
Metro™ Electronic and Cleanroom Shelving Accessory, Label Holder
Conductive label holders
Brand: Metro™ 9990PESD
Additional Details : Weight : 0.00800kg
Description
- For use with Metro™ Super Erecta™ wire shelves and MetroMax Q ESD frames
- Available in two lengths
Specifications
3 in. | |
1.25 in. | |
7.6 cm | |
Plastic |
3.2 cm | |
SmartTray™ system | |
Blue |