Få mere at vide
Metrizamide, TRC
Artikelnummer. 30484105
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30484105 100 mg
30481963 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30484105 Leverandør Toronto Research Chemicals Leverandørnr. TRCM338850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 31112-62-6
Molekylær formel C18 H22 I3 N3 O8
Molekylvægt (g/mol) 789.1
Synonym D-Glucose, 2-[[3-(acetylamino)-5-(acetylmethylamino)-2,4,6-triiodobenzoyl]amino]-2-deoxy-, D-Glucose, 2-[3-acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzamido]-2-deoxy- (8CI), 2-[[3-(Acetylamino)-5-(acetylmethylamino)-2,4,6-triiodobenzoyl]amino]-2-deoxy-D-glucose, Amipaque, Metrizamide, Telebrix 300
IUPAC navn 3-acetamido-5-[acetyl(methyl)amino]-2,4,6-triiodo-N-[(2R,3R,4S,5R)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]benzamide
SMIL CN(C(=O)C)c1c(I)c(NC(=O)C)c(I)c(C(=O)N[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO)c1I
Tekniske data
Kemisk navn eller materiale Metrizamide
Mængde 100 mg
InChI formel InChI=1S/C18H22I3N3O8/c1-6(27)22-14-11(19)10(12(20)15(13(14)21)24(3)7(2)28)18(32)23-8(4-25)16(30)17(31)9(29)5-26/h4,8-9,16-17,26,29-31H,5H2,1-3H3,(H,22,27)(H,23,32)/t8-,9+,16+,17+/m0/s1
Formel vægt 788.8541