Få mere at vide
Metriram (>90%), TRC
Artikelnummer. 30465567
Skift visning
Mængde:
10 g
Artikelnummer. Mængde
30465567 10 g
1 options
Artikelnummer. 30465567 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33863010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metiram
CAS 9006-42-2
Molekylær formel (C16H33N11S16Zn3)x
Mængde 10 g
Synonym Metiram, Polyram, Amarex, Carbatene, Metiram-zinc, NIA 9102, Phytox-Super, Polycarbacin, Polycarbacine, Polycarbazin, Polycarbazine, Polykarbacin, Polymarcin, Polymarcine, Polymarsin, Polymarzin, Polymarzine, Polymat, Polyram 2000, Polyram 80, Polyram 80WP, Polyram DF, Polyram combi
InChI formel InChI=1S/C4H8N2S4.Zn/c7-3(8)5-1-2-6-4(9)10;/h1-2H2,(H2,5,7,8)(H2,6,9,10);/q;+2/p-2
SMIL C(CNC(=S)[S-])NC(=S)[S-].[Zn+2]
IUPAC navn zinc(II) ethane-1,2-diyldicarbamodithioate