Få mere at vide
Metoxuron-d6 (N,N-dimethyl-d6), TRC
Artikelnummer. 30482941
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482941 1 mg
30479052 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482941 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3388441MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metoxuron-d6 (N,N-dimethyl-d6)
CAS 2030182-41-1
Molekylær formel C10 D6 H7 Cl N2 O2
Mængde 1 mg
Synonym Urea, N'-(3-chloro-4-methoxyphenyl)-N,N-di(methyl-d3)- (ACI), N'-(3-Chloro-4-methoxyphenyl)-N,N-di(methyl-d3)urea (ACI), 1-(3-Chloro-4-methoxyphenyl)-3,3-di(methyl-d3)urea, Metoxuron-d6, Metoxuron-D6 (N,N-dimethyl-D6)
InChI formel InChI=1S/C10H13ClN2O2/c1-13(2)10(14)12-7-4-5-9(15-3)8(11)6-7/h4-6H,1-3H3,(H,12,14)/i1D3,2D3
SMIL [2H]C([2H])([2H])N(C(=O)Nc1ccc(OC)c(Cl)c1)C([2H])([2H])[2H]
IUPAC navn 3-(3-chloro-4-methoxyphenyl)-1,1-bis(trideuteriomethyl)urea
Molekylvægt (g/mol) 234.7124
Formel vægt 234.1042