Få mere at vide
Metoxuron-d6 (N,N-dimethyl-d6), CDN
Artikelnummer. 30313828
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313828 0.05 g
30320226 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313828 Leverandør CDN Leverandørnr. CDND76920.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metoxuron-d6 (N,N-dimethyl-d6)
CAS 2030182-41-1
Molekylær formel C10 D6 H7 Cl N2 O2
Mængde 0.05 g
Synonym Urea, N'-(3-chloro-4-methoxyphenyl)-N,N-di(methyl-d3)- (ACI), N'-(3-Chloro-4-methoxyphenyl)-N,N-di(methyl-d3)urea (ACI), 1-(3-Chloro-4-methoxyphenyl)-3,3-di(methyl-d3)urea, Metoxuron-d6, Metoxuron-D6 (N,N-dimethyl-D6)
InChI formel InChI=1 S/C10H13ClN2O2/c1-13(2)10(14)12-7-4-5-9(15-3)8(11)6-7/h4-6 H,1-3H3,(H,12,14)/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])N(C(=O)Nc1ccc(OC)c(Cl)c1)C([2 H])([2 H])[2 H]
IUPAC navn 3-(3-chloro-4-methoxyphenyl)-1,1-bis(trideuteriomethyl)urea
Molekylvægt (g/mol) 234.7124
Formel vægt 234.1042 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity