Få mere at vide
Metoserpate, TRC
Artikelnummer. 17994575
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17994575 1 mg -
17921155 2.5 mg 2.5mg
17902735 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17994575 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3388101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1178-28-5
Molekylær formel C24 H32 N2 O5
Molekylvægt (g/mol) 428.52
Synonym 18-Epireserpic acid, O-methyl-, methyl ester (6CI,7CI), 3β,20α-Yohimban-16β-carboxylic acid, 11,17α,18α-trimethoxy-, methyl ester (8CI), 18-epi-O-Methylreserpic acid methyl ester, Mepiserpate, Methyl O-methyl-18-epi-reserpate, Methyl O-methyl-18-epireserpate, Methyl O-methylepireserpate, Metoserpate, Su 9064
IUPAC navn methyl (1R,15S,17S,18R,19S,20S)-6,17,18-trimethoxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMIL CO[C@H]1C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]([C@H]1OC)C(=O)OC
Tekniske data
Kemisk navn eller materiale Metoserpate
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C24H32N2O5/c1-28-14-5-6-15-16-7-8-26-12-13-9-20(29-2)23(30-3)21(24(27)31-4)17(13)11-19(26)22(16)25-18(15)10-14/h5-6,10,13,17,19-21,23,25H,7-9,11-12H2,1-4H3/t13-,17+,19-,20+,21+,23+/m1/s1
Formel vægt 428.2311
Procent renhed >95%