Få mere at vide
Metoprolol Tartrate EP Impurity M (as Dihydrochloride), Mikromol™
Beskrivelse
Metoprolol Tartrate EP Impurity M (as Dihydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,3-Bis[(1-methylethyl)amino]propan-2-ol Dihydrochloride |
| CAS | 73313-36-7 |
| Analyt- eller komponentnavne | 1,3-Bis[(1-methylethyl)amino]propan-2-ol Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H22 N2 O . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1,3-Bis[(1-methylethyl)amino]propan-2-ol Dihydrochloride, Metoprolol Succinate Imp. M (EP) as Dihydrochloride, Metoprolol Tartrate Imp. M (EP) as Dihydrochloride, Metoprolol Succinate Imp. M (EP), Metoprolol Imp. M (EP), Metoprolol Succinate Impurity M as Dihydrochloride, Metoprolol Tartrate Impurity M as Dihydrochloride, Metoprolol Impurity M as Dihydrochloride, Metoprolol Impurity M as Dihydrochloride |
| InChI formel | InChI=1S/C9H22N2O.2ClH/c1-7(2)10-5-9(12)6-11-8(3)4;;/h7-12H,5-6H2,1-4H3;2*1H |
| SMIL | Cl.Cl.CC(C)NCC(O)CNC(C)C |
| IUPAC navn | 1,3-bis(propan-2-ylamino)propan-2-ol;dihydrochloride |
| Vis mere |