Få mere at vide
Metoprolol Tartrate EP Impurity M (as Dihydrochloride), Mikromol™
Artikelnummer. 30049457
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049457 25 mg
30050264 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049457 Leverandør Mikromol Leverandørnr. MM0027.280025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Metoprolol Tartrate EP Impurity M (as Dihydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Bis[(1-methylethyl)amino]propan-2-ol Dihydrochloride
CAS 73313-36-7
Analyt- eller komponentnavne 1,3-Bis[(1-methylethyl)amino]propan-2-ol Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C9 H22 N2 O . 2 Cl H
Certificeringer/compliance ISO 9001
Synonym 1,3-Bis[(1-methylethyl)amino]propan-2-ol Dihydrochloride, Metoprolol Succinate Imp. M (EP) as Dihydrochloride, Metoprolol Tartrate Imp. M (EP) as Dihydrochloride, Metoprolol Succinate Imp. M (EP), Metoprolol Imp. M (EP), Metoprolol Succinate Impurity M as Dihydrochloride, Metoprolol Tartrate Impurity M as Dihydrochloride, Metoprolol Impurity M as Dihydrochloride, Metoprolol Impurity M as Dihydrochloride
InChI formel InChI=1S/C9H22N2O.2ClH/c1-7(2)10-5-9(12)6-11-8(3)4;;/h7-12H,5-6H2,1-4H3;2*1H
SMIL Cl.Cl.CC(C)NCC(O)CNC(C)C
IUPAC navn 1,3-bis(propan-2-ylamino)propan-2-ol;dihydrochloride
Molekylvægt (g/mol) 247.21
Mængde 25 mg
Formel vægt 246.1266 g/mol