Få mere at vide
Metomidate Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Metomidate Hydrochloride |
| CAS | 35944-74-2 |
| Analyt- eller komponentnavne | Metomidate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H14 N2 O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, methyl ester, hydrochloride (1:1), 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, methyl ester, monohydrochloride (9CI), Metomidate hydrochloride, NSC 168403, Imidazole-5-carboxylic acid, 1-(alpha-methylbenzyl)-, methyl ester, hydrochloride (7CI) |
| InChI formel | InChI=1S/C13H14N2O2.ClH/c1-10(11-6-4-3-5-7-11)15-9-14-8-12(15)13(16)17-2;/h3-10H,1-2H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.COC(=O)c1cncn1C(C)c2ccccc2 |
| Vis mere |