Få mere at vide
Metolachlor D6 (propyl D6), Dr. Ehrenstorfer
Artikelnummer. 30322099
Skift visning
Mængde:
10 mg
Pakningsstørrelse:
10mg
Artikelnummer. Mængde unitSize
30322099 10 mg 10mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322099 Leverandør Dr. Ehrenstorfer Leverandørnr. DRECA15170100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metolachlor D6 (propyl D6)
CAS 1219803-97-0
Molekylær formel C15 2H6 H16 Cl N O2
Mængde 10 mg
Synonym Metolachlor D6 (propyl D6)
InChI formel InChI=1S/C15H22ClNO2/c1-5-13-8-6-7-11(2)15(13)17(14(18)9-16)12(3)10-19-4/h6-8,12H,5,9-10H2,1-4H3/i3D3,10D2,12D
SMIL [2H]C([2H])([2H])C([2H])(N(C(=O)CCl)c1c(C)cccc1CC)C([2H])([2H])OC
IUPAC navn 2-chloro-N-(2-ethyl-6-methylphenyl)-N-(1,1,1,2,3,3-hexadeuterio-3-methoxypropan-2-yl)acetamide
Molekylvægt (g/mol) 289.83
Formel vægt 289.1716 g/mol