Få mere at vide
Metofoline, LoGiCal
Artikelnummer. 30463581
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30463581 5 mg
1 options
Artikelnummer. 30463581 Leverandør LoGiCal Leverandørnr. LGCFOR3218.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metofoline
CAS 2154-02-1
Analyt- eller komponentnavne Metofoline
Forsendelsestilstand Room Temperature
Molekylær formel C20 H24 Cl N O2
Certificeringer/compliance ISO 9001
Synonym Isoquinoline, 1-[2-(4-chlorophenyl)ethyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-, Isoquinoline, 1-(p-chlorophenethyl)-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl- (6CI,7CI,8CI), 1-(p-Chlorophenethyl)-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinoline, 1-(p-Chlorophenethyl)-2-methyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, 1-(p-Chlorophenethyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinoline, ARC 1-K-1, ARC I-K 1, Methopholine, Metofoline, Metopholine, NSC 169889, Ro 4-1778, Versidyne
InChI formel InChI=1S/C20H24ClNO2/c1-22-11-10-15-12-19(23-2)20(24-3)13-17(15)18(22)9-6-14-4-7-16(21)8-5-14/h4-5,7-8,12-13,18H,6,9-11H2,1-3H3
Løsningstype Neat
SMIL COc1cc2CCN(C)C(CCc3ccc(Cl)cc3)c2cc1OC
IUPAC navn 1-[2-(4-chlorophenyl)ethyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinoline
Industry Type Clinical Testing
Molekylvægt (g/mol) 345.86
Mængde 5 mg
Formel vægt 345.1496 g/mol