Få mere at vide
Metoclopramide-d3, TRC
Artikelnummer. 30469278
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30469278 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469278 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3386875MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metoclopramide-d3
CAS 1216522-89-2
Renhedsgrad noter HPLC
Molekylær formel C14 D3 H19 Cl N3 O2
Mængde 5 mg
Synonym 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-(methoxy-d3)benzamide, 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-o-anisamide-d3, 2-(Methoxy-d3)-5-chloroprocainamide, Clopromate-d3, DEL 1267-d3, Draclamid-d3, Emperal-d3, Eucil-d3, Gastrese-d3, Gastrobid-d3, Gastromax-d3, Gastrosil-d3, Gastrotem-d3, Maxeran-d3, Meclopran-d3, Metamide-d3, Reliveran-d3
InChI formel InChI=1S/C14H22ClN3O2/c1-4-18(5-2)7-6-17-14(19)10-8-11(15)12(16)9-13(10)20-3/h8-9H,4-7,16H2,1-3H3,(H,17,19)/i3D3
SMIL [2H]C([2H])([2H])Oc1cc(N)c(Cl)cc1C(=O)NCCN(CC)CC
IUPAC navn 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-(trideuteriomethoxy)benzamide
Molekylvægt (g/mol) 302.8149
Formel vægt 302.1589
Procent renhed >95