Få mere at vide
Metixene Hydrochloride, LoGiCal
Artikelnummer. 30461474
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461474 10 mg
1 options
Artikelnummer. 30461474 Leverandør LoGiCal Leverandørnr. LGCFOR0125.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metixene hydrochloride
CAS 7081-40-5
Analyt- eller komponentnavne Metixene hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C20 H23 N S . Cl H . H2 O
Certificeringer/compliance ISO 9001
Synonym Metixene Hydrochloride, (RS)-1-methyl-3-[(9H-thioxanthen-9-yl)methyl]piperidine hydrochloride
InChI formel InChI=1S/C20H23NS.ClH.H2O/c1-21-12-6-7-15(14-21)13-18-16-8-2-4-10-19(16)22-20-11-5-3-9-17(18)20;;/h2-5,8-11,15,18H,6-7,12-14H2,1H3;1H;1H2
Løsningstype Neat
SMIL O.Cl.CN1CCCC(CC2c3ccccc3Sc4ccccc24)C1
IUPAC navn 1-methyl-3-(9H-thioxanthen-9-ylmethyl)piperidine;hydrate;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 363.94
Mængde 10 mg
Formel vægt 363.1424 g/mol