Få mere at vide
Methylsulfonal, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methylsulfonal |
| CAS | 76-20-0 |
| Analyt- eller komponentnavne | Methylsulfonal |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H18 O4 S2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Butane, 2,2-bis(ethylsulfonyl)-, 2,2-Bis(ethylsulfonyl)butane, Ethylsulfonal, Methylsulfonal, NSC 3994, Sulfonethylmethane, Tional, Trional |
| InChI formel | InChI=1S/C8H18O4S2/c1-5-8(4,13(9,10)6-2)14(11,12)7-3/h5-7H2,1-4H3 |
| Løsningstype | Neat |
| SMIL | CCC(C)(S(=O)(=O)CC)S(=O)(=O)CC |
| Vis mere |