Få mere at vide
Methylprednisolone Hydrogen Succinate Sodium Salt, Mikromol™
Beskrivelse
Methylprednisolone Hydrogen Succinate Sodium Salt
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methylprednisolone Hydrogen Succinate Sodium Salt |
| CAS | 3-3-2375 |
| Analyt- eller komponentnavne | Methylprednisolone Hydrogen Succinate Sodium Salt |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C26 H33 O8 . Na |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Methylprednisolone Hydrogen Succinate Sodium Salt |
| InChI formel | InChI=1S/C26H34O8.Na/c1-14-10-16-17-7-9-26(33,20(29)13-34-22(32)5-4-21(30)31)25(17,3)12-19(28)23(16)24(2)8-6-15(27)11-18(14)24;/h6,8,11,14,16-17,19,23,28,33H,4-5,7,9-10,12-13H2,1-3H3,(H,30,31);/q;+1/p-1/t14-,16-,17-,19-,23+,24-,25-,26-;/m0./s1 |
| SMIL | [Na+].C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)CCC(=O)[O-])[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14 |
| IUPAC navn | sodium;4-[2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoate |
| Vis mere |