Få mere at vide
Methylprednisolone Acetate, Mikromol™
Beskrivelse
Methylprednisolone Acetate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methylprednisolone Acetate |
| CAS | 53-36-1 |
| Analyt- eller komponentnavne | Methylprednisolone Acetate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C24 H32 O6 |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Methylprednisolone Acetate, Methylprednisolone Hydrogen Succinate Imp. C (EP), 6-Methylprednisolone acetate, DepMedalone 40, DepMedalone 80, Depo-Medrate, Depo-Medrol, Depo-Medrone, Depo-methylprednisolone, MPA, Medrol acetate, Mepred, Methylprednisolone 21-acetate, Methylprednisolone acetate, NSC 48985, U 8210, Urbason Crystal Suspension, Vetacortyl, Pregna-1,4-diene-3,20-dione, 11beta,17,21-trihydroxy-6alpha-methyl-, 21-acetate (6CI,7CI,8CI), (6alpha,11beta)-21-(Acetyloxy)-11,17-dihydroxy-6-methylpregna-1,4-diene-3,20-dione, 11beta,17alpha,21-Trihydroxy-6alpha-methylpregna-1,4-diene-3,20-dione 21-acetate, 21-Acetoxy-11beta,17-dihydroxy-6alpha-methylpregna-1,4-diene-3,20-dione, 6alpha-Methylprednisolone 21-acetate, 6alpha-Methylprednisolone acetate, Methylprednisolone Hydrogen Succinate Impurity C, Methylprednisolone Impurity J |
| InChI formel | InChI=1S/C24H32O6/c1-13-9-16-17-6-8-24(29,20(28)12-30-14(2)25)23(17,4)11-19(27)21(16)22(3)7-5-15(26)10-18(13)22/h5,7,10,13,16-17,19,21,27,29H,6,8-9,11-12H2,1-4H3/t13-,16-,17-,19-,21+,22-,23-,24-/m0/s1 |
| SMIL | C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)C)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14 |
| IUPAC navn | [2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Vis mere |