Få mere at vide
Methylphenidate Carboxylic Acid Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ritalinic acid HCl |
| CAS | 19395-40-5 |
| Molekylær formel | C13H17NO2.HCl |
| Mængde | 50 mg |
| Synonym | Ritalinic cid hydrochloride, Methylphenidate Related Compound A (USP), 2-Piperidineacetic acid, α-phenyl-, hydrochloride (1:1) (ACI), 2-Piperidineacetic acid, α-phenyl-, hydrochloride (6CI, 7CI, 8CI, 9CI), 2-Phenyl-2-(piperidin-2-yl)acetic acid hydrochloride, α-Phenyl-2-piperidineacetic acid hydrochloride |
| InChI formel | InChI=1S/C13H17NO2.ClH/c15-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-14-11;/h1-3,6-7,11-12,14H,4-5,8-9H2,(H,15,16);1H |
| SMIL | Cl.OC(=O)C(C1CCCCN1)c2ccccc2 |
| IUPAC navn | 2-phenyl-2-piperidin-2-ylacetic acid;hydrochloride |
| Molekylvægt (g/mol) | 255.74 |
| Formel vægt | 255.1026 |