Få mere at vide
Methylmalonic Acid-d3, TRC
Artikelnummer. 30484230
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30484230 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30484230 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31886210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl-D3-malonic acid
CAS 42522-59-8
Renhedsgrad noter HPLC
Molekylær formel C4 2H3 H3 O4
Mængde 10 mg
Synonym 2-Methylmalonic Acid D3, Propanedioic acid','methyl-d3- (9CI), 2-(Methyl-d3)propanedioic acid
InChI formel InChI=1S/C4H6O4/c1-2(3(5)6)4(7)8/h2H,1H3,(H,5,6)(H,7,8)/i1D3
SMIL [2H]C([2H])([2H])C(C(=O)O)C(=O)O
IUPAC navn 2-(trideuteriomethyl)propanedioic acid
Molekylvægt (g/mol) 121.11
Formel vægt 121.0454
Procent renhed >95