Få mere at vide
Methyleugenol, LoGiCal
Artikelnummer. 30460305
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460305 10 mg
1 options
Artikelnummer. 30460305 Leverandør LoGiCal Leverandørnr. LGCFOR0877.19
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Eugenol Methyl Ether
CAS 93-15-2
Analyt- eller komponentnavne Eugenol Methyl Ether
Forsendelsestilstand Room Temperature
Molekylær formel C11 H14 O2
Certificeringer/compliance ISO 9001
Synonym Benzene, 1,2-dimethoxy-4-(2-propenyl)- (9CI), Benzene, 4-allyl-1,2-dimethoxy- (8CI), Veratrole, 4-allyl- (6CI), 1,2-Dimethoxy-4-(2-propenyl)benzene, 1,2-Dimethoxy-4-allylbenzene, 1,3,4-Eugenol methyl ether, 1-(3,4-Dimethoxyphenyl)-2-propene, 1-Allyl-3,4-dimethoxybenzene, 3,4-Dimethoxy-1-(2-propenyl)benzene, 3,4-Dimethoxyallylbenzene, 3-(3,4-Dimethoxyphenyl)-1-propene, 3-(3,4-Dimethoxyphenyl)propene, 4-Allyl-1,2-dimethoxybenzene, 4-Allylveratrole, Chavibetol methyl ether, Ent 21040, Eugenol methyl ether, Eugenyl methyl ether, Methyl eugenol ether, Methyl eugenyl ether, Methylchavibetol, Methyleugenol, NSC 209528, NSC 8900, O-Methyleugenol, Veratrole methyl ether
InChI formel InChI=1S/C11H14O2/c1-4-5-9-6-7-10(12-2)11(8-9)13-3/h4,6-8H,1,5H2,2-3H3
Løsningstype Neat
SMIL COc1ccc(CC=C)cc1OC
IUPAC navn 1,2-dimethoxy-4-prop-2-enylbenzene
Industry Type Clinical Testing
Molekylvægt (g/mol) 178.23
Mængde 10 mg
Formel vægt 178.0994 g/mol