Learn More
Methyl trifluoroacetate, 99%, Thermo Scientific Chemicals
Catalog Number p-3758507
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
431-47-0
C3H3F3O2
128.05
MFCD00000417
VMVNZNXAVJHNDJ-UHFFFAOYSA-N
methyl trifluoroacetate, trifluoroacetic acid methyl ester, acetic acid, trifluoro-, methyl ester, methyltrifluoroacetate, 2,2,2-trifluoromethylacetate, acetic acid, 2,2,2-trifluoro-, methyl ester, methyl trifluoracetate, tfamt, methyl triflouroacetate, pubchem12490
9893
methyl 2,2,2-trifluoroacetate
COC(=O)C(F)(F)F
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
431-47-0
C3H3F3O2
128.05
MFCD00000417
VMVNZNXAVJHNDJ-UHFFFAOYSA-N
methyl trifluoroacetate, trifluoroacetic acid methyl ester, acetic acid, trifluoro-, methyl ester, methyltrifluoroacetate, 2,2,2-trifluoromethylacetate, acetic acid, 2,2,2-trifluoro-, methyl ester, methyl trifluoracetate, tfamt, methyl triflouroacetate, pubchem12490
9893
methyl 2,2,2-trifluoroacetate
COC(=O)C(F)(F)F
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -78°C
Color Colorless
Density 1.2730g/mL
Boiling Point 43°C to 43.5°C
Flash Point -7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3CO2CH3
Refractive Index 1.2890 to 1.291
Quantity 25 mL
Specific Gravity 1.273
Solubility Information Solubility in water: 8g/L water
Viscosity 0.31 mPa.s (20°C)
Formula Weight 128.05
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl trifluoroacetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl trifluoroacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.