missing translation for 'onlineSavingsMsg'
Få mere at vide

Methylephedrine ((1R,2S)-Methylephedrine), Mikromol™

Artikelnummer. 30050690
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050690 100 mg
30049636 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30050690 Leverandør Mikromol Leverandørnr. MM0652.04

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methylephedrine ((1R,2S)-Methylephedrine)
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methylephedrine
CAS 552-79-4
Analyt- eller komponentnavne Methylephedrine
Forsendelsestilstand Room Temperature
Molekylær formel C11 H17 N O
Certificeringer/compliance ISO 17025
Synonym Methylephedrine, (1R,2S)-Methylephedrine, (-)-Methylephedrine, (-)-N-Methylephedrine, (1R,2S)-(-)-N-Methylephedrine, (1R,2S)-2-Dimethylamino-1-phenyl-1-propanol, (1R,2S)-2-Dimethylamino-1-phenylpropanol, (1R,2S)-3-Dimethylamino-1-phenyl-1-propanol, (1R,2S)-N-Methylephedrin, (1R,2S)-N-Methylephedrine, L-N-Methylephedrine, N-Methyl-L-ephedrine, N-Methylephedrine, (alphaR)-alpha-[(1S)-1-(Dimethylamino)ethyl]benzenemethanol, [R-(R*,S*)]-alpha-[1-(Dimethylamino)ethyl]benzenemethanol, (-)-alpha-[1-(Dimethylamino)ethyl]benzyl alcohol
InChI formel InChI=1S/C11H17NO/c1-9(12(2)3)11(13)10-7-5-4-6-8-10/h4-9,11,13H,1-3H3/t9-,11-/m0/s1
SMIL C[C@@H]([C@H](O)c1ccccc1)N(C)C
IUPAC navn (1R,2S)-2-(dimethylamino)-1-phenylpropan-1-ol
Molekylvægt (g/mol) 179.26
Mængde 100 mg
Formel vægt 179.131 g/mol
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.