missing translation for 'onlineSavingsMsg'
Learn More

Methylene bis-(Chlorosulfate) (>85%), TRC

Product Code. 30482969
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30482969 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482969 Supplier Toronto Research Chemicals Supplier No. TRCM28990510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Methylene bis-(Chlorosulfate)
CAS 92975-18-3
Molekylær formel CH2 Cl2 O6 S2
Mængde 10 mg
Synonym S,S-Methylene Chlorosulfuric Acid Ester
InChI formel InChI=1S/CH2Cl2O6S2/c2-10(4,5)8-1-9-11(3,6)7/h1H2
SMIL ClS(=O)(=O)OCOS(=O)(=O)Cl
IUPAC navn bis(chlorosulfonyloxy)methane
Molekylvægt (g/mol) 245.06
Formel vægt 243.867
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.