missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-chlorophenylacetate, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Color | Colorless to Yellow |
Density | 1.2000g/mL |
Quantity | 25 g |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Physical Form | Liquid |
Refractive Index | 1.5225 to 1.5245 |
Assay Percent Range | 97.5% min. (GC) |
Specific Gravity | 1.2 |
Linear Formula | ClC6H4CH2C(O)OCH3 |
Show More |
Safety and Handling
Product Identifier
- Methyl 3-chlorophenylacetate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'